Assay Details
Gene Symbol: STOX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001491926.6 | XP_001491976.1 | 2007 | 85 | ||
XM_014736634.2 | XP_014592120.1 | 1531 | 85 | |||
XM_023630417.1 | XP_023486185.1 | 1544 | 85 | |||
XM_023630418.1 | XP_023486186.1 | 1752 | 85 | |||
XM_023630419.1 | XP_023486187.1 | 1544 | 85 | |||
XM_023630420.1 | XP_023486188.1 | 1545 | 85 | |||
XM_023630421.1 | XP_023486189.1 | 627 | 85 | |||
XM_023630422.1 | XP_023486190.1 | 1356 | 85 | |||
XM_023630423.1 | XP_023486191.1 | 362 | 85 | |||
XM_023630424.1 | XP_023486192.1 | 1279 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
