Assay Details
Gene Symbol: RWDD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001492050.4 | XP_001492100.1 | 695 | 87 | ||
XM_005606403.3 | XP_005606460.1 | 696 | 87 | |||
XM_005606404.3 | XP_005606461.1 | 697 | 87 | |||
XM_014736636.2 | XP_014592122.1 | 451 | 87 | |||
XM_023630585.1 | XP_023486353.1 | 697 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
