Assay Details
Gene Symbol: CDKN2AIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023630883.1 | XP_023486651.1 | 446 | 79 | ||
XM_023630884.1 | XP_023486652.1 | 573 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
