Assay Details
Gene Symbol: RAB21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023631458.1 | XP_023487226.1 | 466 | 69 | ||
GenBank mRNA | JL616024.1 | - | 2 - 3 | 104 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
