Assay Details
Gene Symbol: CSRP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023631768.1 | XP_023487536.1 | 417 | 59 | ||
XM_023631770.1 | XP_023487538.1 | 574 | 59 | |||
XM_023631771.1 | XP_023487539.1 | 412 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
