Assay Details
Gene Symbol: PAWR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023631519.1 | XP_023487287.1 | 1185 | 85 | ||
XM_023631520.1 | XP_023487288.1 | 1196 | 85 | |||
XM_023631521.1 | XP_023487289.1 | 1071 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
