Assay Details
Gene Symbol: MRPL42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014736747.2 | XP_014592233.1 | 318 | 68 | ||
XM_023631866.1 | XP_023487634.1 | 320 | 68 | |||
XM_023631867.1 | XP_023487635.1 | 231 | 68 | |||
XM_023631868.1 | XP_023487636.1 | 320 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
