Assay Details
Gene Symbol: CRADD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005606505.3 | XP_005606562.1 | 777 | 110 | ||
XM_023631423.1 | XP_023487191.1 | 1024 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
