Assay Details
Gene Symbol: GAS2L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023631175.1 | XP_023486943.1 | 363 | 110 | ||
XM_023631176.1 | XP_023486944.1 | 3465 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
