Assay Details
Gene Symbol: GLT8D2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001498127.6 | XP_001498177.1 | 1184 | 61 | ||
XM_005606556.3 | XP_005606613.1 | 1286 | 61 | |||
XM_014736777.2 | XP_014592263.1 | 819 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
