Assay Details
Gene Symbol: C1QTNF6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499418.5 | XP_001499468.3 | 155 | 58 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001499418.5 transcripts.
Set Membership: |
![]() ![]() |
