Assay Details
Gene Symbol: DNAL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005606645.3 | XP_005606702.1 | 312 | 85 | ||
XM_023631186.1 | XP_023486954.1 | 686 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
