Assay Details
Gene Symbol: CENPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001500392.5 | XP_001500442.3 | 409 | 70 | ||
XM_023631044.1 | XP_023486812.1 | 316 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
