Assay Details
Gene Symbol: TSPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001503143.5 | XP_001503193.1 | 481 | 52 | ||
XM_005606712.3 | XP_005606769.1 | 610 | 52 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
