Assay Details
Gene Symbol: PRR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023631383.1 | XP_023487151.1 | 947 | 78 | ||
XM_023631384.1 | XP_023487152.1 | 776 | 78 | |||
XM_023631385.1 | XP_023487153.1 | 751 | 78 | |||
XM_023631386.1 | XP_023487154.1 | 786 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
