Assay Details
Gene Symbol: ARHGAP8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014736939.2 | XP_014592425.2 | 1248 | 58 | ||
XM_023631375.1 | XP_023487143.1 | 1659 | 58 | |||
XM_023631376.1 | XP_023487144.1 | 1659 | 58 | |||
XM_023631377.1 | XP_023487145.1 | 1073 | 58 | |||
XM_023631379.1 | XP_023487147.1 | 962 | 58 | |||
XM_023631380.1 | XP_023487148.1 | 858 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
