Assay Details
Gene Symbol: TTLL8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023631750.1 | XP_023487518.1 | 875 | 56 | ||
XM_023631751.1 | XP_023487519.1 | 874 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() |
