Assay Details
Gene Symbol: MAPK11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001490089.5 | XP_001490139.3 | 805 | 60 | ||
XM_005606799.2 | XP_005606856.2 | 812 | 60 | |||
XM_005606800.3 | XP_005606857.1 | 834 | 60 | |||
XM_023631321.1 | XP_023487089.1 | 778 | 60 | |||
XM_023631322.1 | XP_023487090.1 | 657 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
