Assay Details
Gene Symbol: TYMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023631572.1 | XP_023487340.1 | 947 | 59 | ||
XM_023631573.1 | XP_023487341.1 | 773 | 59 | |||
XM_023631574.1 | XP_023487342.1 | 943 | 59 | |||
XM_023631575.1 | XP_023487343.1 | 942 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
