Assay Details
Gene Symbol: CHKB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023631564.1 | XP_023487332.1 | 951 | 70 | ||
XM_023631565.1 | XP_023487333.1 | 951 | 70 | |||
XM_023631566.1 | XP_023487334.1 | 774 | 70 | |||
XM_023631567.1 | XP_023487335.1 | 774 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
