Assay Details
Gene Symbol: ENKUR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005606883.3 | XP_005606940.1 | 416 | 102 | ||
XM_005606884.3 | XP_005606941.1 | 322 | 102 | |||
XM_014737036.2 | XP_014592522.1 | 419 | 102 | |||
XM_023632320.1 | XP_023488088.1 | 555 | 102 | |||
XM_023632321.1 | XP_023488089.1 | 420 | 102 | |||
XM_023632322.1 | XP_023488090.1 | 425 | 102 | |||
XM_023632323.1 | XP_023488091.1 | 238 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
