Assay Details
Gene Symbol: PRTFDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023632392.1 | XP_023488160.1 | 561 | 67 | ||
XM_023632393.1 | XP_023488161.1 | 574 | 67 | |||
XM_023632394.1 | XP_023488162.1 | 765 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
