Assay Details
Gene Symbol: LOC102148742 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005606891.3 | XP_005606948.1 | 315 | 98 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005606891.3 transcripts.
Set Membership: |
![]() ![]() |
