Assay Details
Gene Symbol: LOC102148742 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005606891.3 | XP_005606948.1 | 659 | 74 | ||
XM_005606892.3 | XP_005606949.1 | 413 | 74 | |||
XM_005606893.3 | XP_005606950.1 | 365 | 74 | |||
XM_023632367.1 | XP_023488135.1 | 813 | 74 | |||
XM_023632368.1 | XP_023488136.1 | 363 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() |
