Assay Details
Gene Symbol: LOC102148742 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005606891.3 | XP_005606948.1 | 755 | 132 | ||
XM_005606892.3 | XP_005606949.1 | 509 | 132 | |||
XM_005606893.3 | XP_005606950.1 | 461 | 132 | |||
XM_023632367.1 | XP_023488135.1 | 909 | 132 | |||
XM_023632368.1 | XP_023488136.1 | 459 | 132 |

More Information
Set Membership: |
![]() ![]() |
