Assay Details
Gene Symbol: MLLT10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014737059.2 | XP_014592545.2 | 1045 | 98 | ||
XM_023632280.1 | XP_023488048.1 | 1145 | 98 | |||
XM_023632281.1 | XP_023488049.1 | 879 | 98 | |||
XM_023632282.1 | XP_023488050.1 | 1017 | 98 | |||
XM_023632283.1 | XP_023488051.1 | 1145 | 98 | |||
XM_023632284.1 | XP_023488052.1 | 1145 | 98 | |||
XM_023632285.1 | XP_023488053.1 | 1145 | 98 | |||
XM_023632286.1 | XP_023488054.1 | 1145 | 98 | |||
XM_023632288.1 | XP_023488056.1 | 1045 | 98 | |||
XM_023632289.1 | XP_023488057.1 | 779 | 98 | |||
XM_023632290.1 | XP_023488058.1 | 1145 | 98 | |||
XM_023632291.1 | XP_023488059.1 | 699 | 98 | |||
XM_023632293.1 | XP_023488061.1 | 714 | 98 | |||
XM_023632294.1 | XP_023488062.1 | 595 | 98 | |||
XM_023632295.1 | XP_023488063.1 | 1145 | 98 | |||
XM_023632296.1 | XP_023488064.1 | 1145 | 98 | |||
XM_023632297.1 | XP_023488065.1 | 1145 | 98 | |||
XM_023632298.1 | XP_023488066.1 | 1145 | 98 | |||
XM_023632299.1 | XP_023488067.1 | 1144 | 98 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
