Assay Details
Gene Symbol: HACD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023632105.1 | XP_023487873.1 | 426 | 72 | ||
XM_023632106.1 | XP_023487874.1 | 313 | 72 | |||
XM_023632107.1 | XP_023487875.1 | 280 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
