Assay Details
Gene Symbol: SUV39H2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005606910.3 | XP_005606967.3 | 1720 | 75 | ||
XM_023632000.1 | XP_023487768.1 | 1268 | 75 | |||
XM_023632001.1 | XP_023487769.1 | 1137 | 75 | |||
GenBank mRNA | JL625103.1 | - | 3 - 4 | 447 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
