Assay Details
Gene Symbol: ASB13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005606955.3 | XP_005607012.1 | 715 | 94 | ||
XM_023632342.1 | XP_023488110.1 | 491 | 94 | |||
XM_023632343.1 | XP_023488111.1 | 723 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
