Assay Details
Gene Symbol: TENT4B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023636936.1 | XP_023492704.1 | 758 | 110 | ||
XM_023636937.1 | XP_023492705.1 | 758 | 110 |

More Information
Set Membership: |
![]() ![]() |
