Assay Details
Gene Symbol: CYLD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023636962.1 | XP_023492730.1 | 2993 | 71 | ||
XM_023636963.1 | XP_023492731.1 | 3014 | 71 | |||
XM_023636965.1 | XP_023492733.1 | 2680 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
