Assay Details
Gene Symbol: USB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001494681.4 | XP_001494731.1 | 356 | 73 | ||
XM_003364612.4 | XP_003364660.1 | 358 | 73 | |||
XM_005608294.3 | XP_005608351.1 | 356 | 73 | |||
XM_005608295.3 | XP_005608352.1 | 356 | 73 | |||
XM_023637089.1 | XP_023492857.1 | 355 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
