Assay Details
Gene Symbol: CKLF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003364591.4 | XP_003364639.1 | 294 | 81 | ||
XM_005608308.3 | XP_005608365.1 | 448 | 81 | |||
XM_023637130.1 | XP_023492898.1 | 500 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
