Assay Details
Gene Symbol: CMTM3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023637132.1 | XP_023492900.1 | 600 | 102 | ||
XM_023637133.1 | XP_023492901.1 | 684 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
