Assay Details
Gene Symbol: GFOD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001915947.5 | XP_001915982.1 | 526 | 70 | ||
XM_005608338.3 | XP_005608395.1 | 885 | 70 | |||
XM_005608339.3 | XP_005608396.1 | 880 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
