Assay Details
Gene Symbol: SLC7A6OS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001499093.5 | XP_001499143.2 | 715 | 63 | ||
GenBank mRNA | JL622276.1 | - | 1 - 2 | 165 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
