Assay Details
Gene Symbol: IST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001916072.5 | XP_001916107.2 | 419 | 66 | ||
XM_005608384.3 | XP_005608441.1 | 419 | 66 | |||
XM_005608385.3 | XP_005608442.1 | 419 | 66 | |||
XM_023637251.1 | XP_023493019.1 | 304 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
