Assay Details
Gene Symbol: FCSK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014738342.2 | XP_014593828.1 | 907 | 72 | ||
XM_023637309.1 | XP_023493077.1 | 899 | 72 | |||
XM_023637310.1 | XP_023493078.1 | 849 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
