Assay Details
Gene Symbol: BCAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005608435.3 | XP_005608492.1 | 801 | 111 | ||
XM_014738367.2 | XP_014593853.1 | 778 | 111 | |||
XM_023637340.1 | XP_023493108.1 | 877 | 111 |

More Information
Set Membership: |
![]() ![]() |
