Assay Details
Gene Symbol: BCAR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005608435.3 | XP_005608492.1 | 1080 | 65 | ||
XM_014738367.2 | XP_014593853.1 | 1057 | 65 | |||
XM_014738368.2 | XP_014593854.1 | 5411 | 65 | |||
XM_023637340.1 | XP_023493108.1 | 1156 | 65 | |||
XM_023637341.1 | XP_023493109.1 | 470 | 65 | |||
XM_023637342.1 | XP_023493110.1 | 445 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
