Assay Details
Gene Symbol: ATMIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023637367.1 | XP_023493135.1 | 470 | 96 | ||
XM_023637368.1 | XP_023493136.1 | 1015 | 96 | |||
XM_023637370.1 | XP_023493138.1 | 432 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023637368.1, XM_023637370.1 transcripts.
Set Membership: |
![]() ![]() |
