Assay Details
Gene Symbol: ATMIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023637367.1 | XP_023493135.1 | 667 | 85 | ||
XM_023637368.1 | XP_023493136.1 | 1212 | 85 | |||
XM_023637370.1 | XP_023493138.1 | 629 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
