Assay Details
Gene Symbol: ADAD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_014738503.2 | XP_014593989.1 | 467 | 70 | ||
XM_023637408.1 | XP_023493176.1 | 468 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
