Assay Details
Gene Symbol: CTDSP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023643898.1 | XP_023499666.1 | 873 | 65 | ||
XM_023643899.1 | XP_023499667.1 | 837 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
