Assay Details
Gene Symbol: MSRB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023643953.1 | XP_023499721.1 | 493 | 91 | ||
XM_023643955.1 | XP_023499723.1 | 504 | 91 | |||
XM_023643956.1 | XP_023499724.1 | 459 | 91 | |||
XM_023643957.1 | XP_023499725.1 | 623 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
