Assay Details
Gene Symbol: CPM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001917056.4 | XP_001917091.2 | 679 | 61 | ||
XM_023643978.1 | XP_023499746.1 | 679 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
