Assay Details
Gene Symbol: YEATS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023643986.1 | XP_023499754.1 | 568 | 122 | ||
XM_023643987.1 | XP_023499755.1 | 568 | 122 | |||
GenBank mRNA | JL619837.1 | - | 5 - 6 | 438 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
