Assay Details
Gene Symbol: PWWP3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023646297.1 | XP_023502065.1 | 765 | 98 | ||
XM_023646298.1 | XP_023502066.1 | 765 | 98 | |||
XM_023646299.1 | XP_023502067.1 | 130 | 98 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_023646297.1, XM_023646298.1, XM_023646299.1 transcripts.
Set Membership: |
![]() ![]() |
