Assay Details
Gene Symbol: MOB3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001504389.5 | XP_001504439.1 | 897 | 123 | ||
XM_005611449.3 | XP_005611506.1 | 898 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() |
