Assay Details
Gene Symbol: CHMP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_023637543.1 | XP_023493311.1 | 220 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
